Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, For 29/26-29/42 Outer Joints, White
SDP
Catalog No. CG302230
Encompass_Preferred
Septum Stopper, Sleeve Type, For 29/26-29/42 Outer Joints, White. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Catalog No. CG302230 Supplier Chemglass Life Sciences Supplier No. CG302230
Septum Stopper, Sleeve Type, For 29/26-29/42 Outer Joints, White. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.