Learn More
Andwin Scientific WHT TOUGH-TAG MICRO.TUBE LABEL
SDP
Supplier:  Andwin Scientific TTLW1000
Encompass_Preferred
The Tough-Tags feature resistance against chemicals and capacity to work within the temperature range of -40C to 121C without peeling, cracking, or falling off. The labels will not jam in laser printers. This product comes with guidelines for printing with Microsoft Word. The labels strongly stick onto several surfaces like plastics and other materials and measure 1.28" x 0.50". It is sold in the color white.
Catalog No. NC9634357