Learn More
Electronic Imaging Materials Inc 651 XyResist CytoLabel 0.88 inch x 0.88 inch White Plastic Slide Labels, Labels are 3 across, 6,600 labels per roll, 1 inch core, 3 ROLLS
SDP
Catalog No. NC0632444
Encompass_Preferred
651—0.88 inch x 0.88 inch XyResist CytoLabel is ideal for printing slide labels right in your Cytopathology or Histology lab. Designed for durable thermal transfer printing, XyResist labels offer excellent print quality and will withstand repeated handling in harsh solvents and chemicals such as DMSO, Isopropyl Alcohol (IPA) and Xylene. The premium adhesive handles cold temperatures well. Works best with T44 premium resin ribbon. For other quantities, please contact customer service. *NOTE: for staining applications, use 626 XyResist chemical-resistant polyester labels.
Catalog No. NC0632444 Supplier Electronic Imaging Materials Inc Supplier No. 6510880881 ROLL TIER
651—0.88 inch x 0.88 inch XyResist CytoLabel is ideal for printing slide labels right in your Cytopathology or Histology lab. Designed for durable thermal transfer printing, XyResist labels offer excellent print quality and will withstand repeated handling in harsh solvents and chemicals such as DMSO, Isopropyl Alcohol (IPA) and Xylene. The premium adhesive handles cold temperatures well. Works best with T44 premium resin ribbon. For other quantities, please contact customer service. *NOTE: for staining applications, use 626 XyResist chemical-resistant polyester labels.