Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Why Do Doctors Test Urine?
SDPSCI_ED

Catalog No. S97678 Shop All EDVOTEK Products
Change view
Click to view available options
Why Do Doctors Test Urine kit?
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S97678 Why Do Doctors Test Urine kit?
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S97678 Supplier Edvotek Supplier No. DD235

Includes

Patient and control simulated urine samples, chemicals and dipsticks to perform color, pH, glucose and protein tests, content background, experiment procedures, safety information and assessment questions and answers

Painless and non-invasive method to know about our health and diseases.

  • Simple, cost effective and highly informative test
  • Simulated patient urine samples, chemically prepared will be analyzed for color, pH, glucose, hemoglobin and protein levels
  • Results obtained will be correlated to human disease

Order Info

Components for 6 experiments.

Specifications

Product Type Educational Kit
For Use With (Application) Simulated patient urine samples are analyzed
Includes Patient and control simulated urine samples, chemicals and dipsticks to perform color, ph, glucose and protein tests, content background, experiment procedures, safety information and assessment questions and answers
Quantity 6 experiments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.