Learn More
Cole-Parmer WI-FI DATA LOGGING REF/FRZ
Catalog No. NC1924809 Shop All Cole-Parmer Products
Encompass_Preferred
Traceable Wi-Fi Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE Cloud Service; 2 Bullet Probes.Stay connected to your critical environments wherever you go!Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability*View current temperature conditions anywherecloud-based interface sends email, text, and push notificationsNo software requiredweb, Android, Apple iOS data visibilityIncludes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Catalog No. NC1924809 Supplier Cole-Parmer Supplier No. 9946010
Traceable Wi-Fi Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE Cloud Service; 2 Bullet Probes.Stay connected to your critical environments wherever you go!Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability*View current temperature conditions anywherecloud-based interface sends email, text, and push notificationsNo software requiredweb, Android, Apple iOS data visibilityIncludes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory