Learn More
Electron Microscopy Sciences Visio-Tek™ Digital Wi-Fi/USB Microscope with Long Working Distance, 10x ~ 160x magnification
SDP
Catalog No. 501930500
Encompass_Preferred
Visio-tek digital portable imaging solutions utilize both Wi-Fi and USB transmission and can be connected to iOS and Android smartphones or tablets via the dedicated Visio-tek App, as well as to Windows or Mac computers or laptops through a Wi-Fi/USB connection. Using theVisio-tek free software and application, you can capture, store pictures and video, and take measurements. Flexible, cost effective and user-friendly. Visio-tek microscopes allow Wi-Fi inspections anywhere an image magnification is required. Designed for applications where increased clearance is beneficial while working under magnification. Viewing at a greater magnification also provides a wider viewing angle.
Magnification: 10x ~ 160x
Focus range: 8 ~ 450 mm
Light source: 8 White LEDs
Catalog No. 50-193-0500 Supplier Electron Microscopy Sciences Supplier No. 92107
Visio-tek digital portable imaging solutions utilize both Wi-Fi and USB transmission and can be connected to iOS and Android smartphones or tablets via the dedicated Visio-tek App, as well as to Windows or Mac computers or laptops through a Wi-Fi/USB connection. Using theVisio-tek free software and application, you can capture, store pictures and video, and take measurements. Flexible, cost effective and user-friendly. Visio-tek microscopes allow Wi-Fi inspections anywhere an image magnification is required. Designed for applications where increased clearance is beneficial while working under magnification. Viewing at a greater magnification also provides a wider viewing angle.
Magnification: 10x ~ 160x
Focus range: 8 ~ 450 mm
Light source: 8 White LEDs