Learn More
ATCC WI-38; Lung Fibroblast; Human
Catalog No. 502382882
Encompass
WI-38 cell line is the first human diploid cell line to be used in human vaccine preparation. WI-38 cells were isolated from the lung tissue of a 3-month-old, female, embryo. WI-38 is used in viruscide testing.
Catalog No. 50-238-2882 Supplier ATCC Supplier No. CCL75
Add to Cart
Add to Cart
WI-38 cell line is the first human diploid cell line to be used in human vaccine preparation. WI-38 cells were isolated from the lung tissue of a 3-month-old, female, embryo. WI-38 is used in viruscide testing.