Learn More
Aviva Systems Biology Corp PYM1 Antibody - C-terminal region (ARP60533_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60533_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WIBG. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human WIBG. Alternate names: MGC13064, PYMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-287-077