Learn More
Erdco Engineering Corporation WICKS ERDCO 10/PK
SDP
Catalog No. EXX2L05513
Encompass
Specification Grade Wicks for Smoke Point test consistancy. Extracted & shipped in a desiccated container. Ten individual wicks per package. ASTM D1322, ASTM D1740, IP 57 & ISO 3014
Catalog No. EXX2L05513 Supplier Erdco Engineering Corporation Supplier No. 2L 05513
Specification Grade Wicks for Smoke Point test consistancy. Extracted & shipped in a desiccated container. Ten individual wicks per package. ASTM D1322, ASTM D1740, IP 57 & ISO 3014