Learn More
MSC Fibre-Metal Wide View Visor, Dark Green, Propionate, 8 in. x 16.5 in., 50/Pk.
Catalog No. 5000507097
Encompass
This wide view visor is designed for reliable protection in industrial environments, such as metal fabrication, automotive repair, and construction. Constructed from robust, dark green propionate, it provides clear visibility while shielding against flying debris and hazardous particles. It is compatible with specific mounting crowns for easy installation and a precise fit.
  • Manufactured from durable propionate
  • Dark green lens for glare reduction
  • Compatible with F400 and F500 mounting crowns
  • Meets ANSI Z87.1+ and CSA Z94.3 safety standards
  • Dimensions of 8 in. (height) x 16.5 in. (width) x 0.06 in. (thickness)
  • Suitable for use with other personal protective equipment
Catalog No. 50-005-07097 Supplier MSC Supplier No. 92144310
This wide view visor is designed for reliable protection in industrial environments, such as metal fabrication, automotive repair, and construction. Constructed from robust, dark green propionate, it provides clear visibility while shielding against flying debris and hazardous particles. It is compatible with specific mounting crowns for easy installation and a precise fit.
  • Manufactured from durable propionate
  • Dark green lens for glare reduction
  • Compatible with F400 and F500 mounting crowns
  • Meets ANSI Z87.1+ and CSA Z94.3 safety standards
  • Dimensions of 8 in. (height) x 16.5 in. (width) x 0.06 in. (thickness)
  • Suitable for use with other personal protective equipment