Learn More
Bio Rad Laboratories WIDE MINI-SUBCELL SYSTEM
Catalog No. NC9573587
Encompass
Wide Mini-Sub Cell GT System, With 15 x 10cm tray, and mini-gel caster, Contains a wide platform that can separate 30 samples per comb, or up to 60 per gel, Suited for multiple sample, rapid screening applications
Catalog No. NC9573587 Supplier Bio Rad Laboratories Supplier No. 1704469
Wide Mini-Sub Cell GT System, With 15 x 10cm tray, and mini-gel caster, Contains a wide platform that can separate 30 samples per comb, or up to 60 per gel, Suited for multiple sample, rapid screening applications