Learn More
Hoefer, Inc Hoefer™ SE640 Wide-Mini Vertical Protein Electrophoresis Unit (Complete unit)
SDP
Catalog No. NC2740649
Encompass_Preferred
Extra wide format ideal for quick high-throughput runsFeatures:Maximum glass plate size 18 x 8 cmPermits rapid screening of up to 112 samples at one timeDesign permits gels to be surrounded uniformly by buffer so heat dissipation is efficient. Bands are sharp and lanes are straightAccommodates denaturing or native polyacrylamide gelsRun up to four second dimension gels using IEF strips or tube gelsComplete Unit Includes:Buffer Chamber; Safety Lid with High Voltage Leads; Glass Plates 18 x 8 cm, 6 pcs; Combs, 15-well, 1.5 mm thick, 2 pcs; Spacers, 1.5 mm Thick, 2 cm wide, 4 pcs; Dual Gel CasterBasic Unit Includes:The same as complete unit(SE640-15-1.5) without the combs and spacers. Specifications:Glass Plate Size (W x H) 18 x 8 cmSpacers 0.75 mm, 1.0 mm, 1.5 mmMaximum Power Settings 1000 V, 500 mA, and 50 WMaximum Temperature 45°CIndoor Use 4 to 40°CHumidity Up to 80%Unit Dimensions (W x H x D) 32 x 29 x 14 cm
Catalog No. NC2740649 Supplier Hoefer, Inc Supplier No. SE640151.5
Extra wide format ideal for quick high-throughput runsFeatures:Maximum glass plate size 18 x 8 cmPermits rapid screening of up to 112 samples at one timeDesign permits gels to be surrounded uniformly by buffer so heat dissipation is efficient. Bands are sharp and lanes are straightAccommodates denaturing or native polyacrylamide gelsRun up to four second dimension gels using IEF strips or tube gelsComplete Unit Includes:Buffer Chamber; Safety Lid with High Voltage Leads; Glass Plates 18 x 8 cm, 6 pcs; Combs, 15-well, 1.5 mm thick, 2 pcs; Spacers, 1.5 mm Thick, 2 cm wide, 4 pcs; Dual Gel CasterBasic Unit Includes:The same as complete unit(SE640-15-1.5) without the combs and spacers. Specifications:Glass Plate Size (W x H) 18 x 8 cmSpacers 0.75 mm, 1.0 mm, 1.5 mmMaximum Power Settings 1000 V, 500 mA, and 50 WMaximum Temperature 45°CIndoor Use 4 to 40°CHumidity Up to 80%Unit Dimensions (W x H x D) 32 x 29 x 14 cm