Learn More
Lakeside Manufacturing Inc Wide Mobile Storage Sloped Top Cabinet with Pegboard
Catalog No. 502884425
Encompass_Preferred
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside ST-4879PB Wide Mobile Storage Sloped Top Cabinet has a pegboard. Durable construction with stainless steel body, top, bottom, and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter-weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Made in the USA.
Catalog No. 50-288-4425 Supplier Lakeside Manufacturing Inc Supplier No. ST4879PB
Add to Cart
Add to Cart
Request a Quote
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside ST-4879PB Wide Mobile Storage Sloped Top Cabinet has a pegboard. Durable construction with stainless steel body, top, bottom, and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter-weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Made in the USA.