Learn More
Ace Glass, Inc. Bottle, GLS80 wide neck, 5 liter, clear, Duran 1113949, cs/1
SDP
Catalog No. NC0835665
Encompass
Duran GLS80 wide neck 5L clear lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.
Catalog No. NC0835665 Supplier Ace Glass, Inc. Supplier No. 555909
Duran GLS80 wide neck 5L clear lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.