Learn More
TELEDYNE LEEMAN LABS STANDARD BOTTLE WIDE MOUTH-WITH CAP 250 ML LOW-DENSITY POLYETHYLENE-5 EACH
Catalog No. NC2819141
Encompass
5 Each250 mLCompatible with adapter plate SP7639, SP7640Compatible with XLR-8, EXR-8, ASX-520, ASX-260, XLR-860, ASX-280 and ASX-560
Catalog No. NC2819141 Supplier TELEDYNE LEEMAN LABS Supplier No. SP5228
5 Each250 mLCompatible with adapter plate SP7639, SP7640Compatible with XLR-8, EXR-8, ASX-520, ASX-260, XLR-860, ASX-280 and ASX-560