Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ WHEATON™ Wide Mouth Bottle, Clear
SDP

Catalog No. 06450309 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
Closure Liner:
Foil
Rubber
Quantity:
24/Case
48/Case
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Liner Quantity
06-450-309 500 mL Rubber 24/Case
06-450-306 60 mL Foil 48/Case
06-450-307 125 mL Foil 24/Case
06-450-308 250 mL Foil 48/Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-450-309 Supplier DWK LIFE SCIENCES™ Supplier No. W217973
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wide Mouth Bottle, Clear is a wide mouth volumetric bottle made of clear Type III soda-lime glass with black phenolic screw caps lined with foil or rubber, ideal for general small to large volume storage.

  • Clear, Type III soda-lime glass construction
  • Wide mouth opening for easy filling and dispensing
  • Black phenolic screw caps available with foil or rubber lining
  • Suitable for general laboratory storage of small to large volumes
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Clear
Material Borosilicate Glass (Type 3)
Quantity 24/Case
Sterility Non-sterile
Closure Color Black
Graduations Yes
Closure Included Yes
Graduated Yes
Mouth Wide
Closure Liner Rubber
Closure Material Phenolic
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape Round
Product Type Graduated Volumetric Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.