Learn More
DWK Life Sciences Wheaton™ Wide Mouth Bottle, Safety Coated
SDP
Catalog No. 02927155 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Liner:
Polyethylene
Polyvinyl
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Liner
02-927-155 250 mL Polyethylene
02-927-156 500 mL Polyethylene
02-914-000 500 mL Polyvinyl
02-927-157 1000 mL Polyethylene
06-450-300 1000 mL Polyvinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-927-155 Supplier DWK Life Sciences Supplier No. W217925
Only null left
Add to Cart
Add to Cart
Wide mouth safety coated clear glass bottles with attached screw caps.
  • Clear, Type III soda-lime glass
  • Largest bottles available
  • Ideal for general, small to large volume storage
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Coating Safety Coated
Color Clear
Material Borosilicate Glass (Type 3)
Quantity 12/Case
Closure Color White
Product Type Wide Mouth Bottle
Closure Liner Polyethylene
Closure Material Polypropylene