Learn More
Electron Microscopy Sciences Wide Mouth Cryogenic Vial
SDP
Catalog No. 501899054
Encompass_Preferred
A wide mouth vial with 15 ml capacity. Useful for the storage and retrieval of solid specimens, such as bone, tumor, and organ samples. Made from polypropylene with high- density polyethylene screw closures. Selfstanding. Radiation sterilized, noncytotoxic, non-pyrogenic.
Catalog No. 50-189-9054 Supplier Electron Microscopy Sciences Supplier No. 6185015
A wide mouth vial with 15 ml capacity. Useful for the storage and retrieval of solid specimens, such as bone, tumor, and organ samples. Made from polypropylene with high- density polyethylene screw closures. Selfstanding. Radiation sterilized, noncytotoxic, non-pyrogenic.