Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ WHEATON™ Wide Mouth Packer, Amber
SDP

Catalog No. 02912399 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1250 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-912-399 2500 mL
02-912-398 1250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-912-399 Supplier DWK LIFE SCIENCES™ Supplier No. W217932
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wide Mouth Packer, Amber bottles are made of amber USP Type III soda-lime glass with an attached white polypropylene PTFE-lined cap, ideal for storing light-sensitive samples or dry powders in field or laboratory settings.

  • Amber, USP Type III soda-lime glass
  • Ideal for storage of light-sensitive samples or dry powders
  • Wide-mouth design for easy addition and removal of contents
  • Attached white polypropylene cap with PTFE-faced liner
  • Suitable for field or laboratory use
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2500 mL
Color Amber
Material Soda Lime Glass
Sterility Non-sterile
Closure Color White
Closure Included Yes
Graduated No
Mouth Wide
Closure Liner PTFE
Closure Material Polypropylene
Autoclavable Not Autoclavable
Capacity (English) 85 oz.
Shape Round
Product Type Wide Mouth Packer Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.