Learn More
Research Products International Corp Wide-Mouth Copolymer Plastic Scintillation Vials, 500 per Case
SDP
Catalog No. 50212996
Encompass
Wide-mouth opening (23mm) for large samples such as filter discs. Constructed of translucent copolymer polypropylene with exceptional clarity. Supplied with a white polyethylene cap that seals with a 1/4 turn. Supplied as bulk bag of vials and bag of caps.
Catalog No. 50-212-996 Supplier Research Products International Corp Supplier No. 121039
Wide-mouth opening (23mm) for large samples such as filter discs. Constructed of translucent copolymer polypropylene with exceptional clarity. Supplied with a white polyethylene cap that seals with a 1/4 turn. Supplied as bulk bag of vials and bag of caps.