Learn More
Chemglass Life Sciences Bottle, Wide Mouth, Amber, Convenience Pack, 28-400 Thread Size, 30mL/1oz, PTFE Lined Green Cap
SDP
Catalog No. NC1214762
Encompass_Preferred
Bottle, Wide Mouth, Amber, Convenience Pack, 30mL/1oz, PTFE Lined Green Cap
Catalog No. NC1214762 Supplier Chemglass Life Sciences Supplier No. CG82103
Bottle, Wide Mouth, Amber, Convenience Pack, 30mL/1oz, PTFE Lined Green Cap