Learn More
BURKLE INC Sealable wide-necked bottle, HDPE, 33.81 oz. (1000 ml), with cap
Catalog No. 5000025806
Encompass_Preferred
The sealable wide-neck bottle protects against unauthorized access or tampering. Sealing eyelets on the cap and bottle can be used to ensure that the bottle has not been opened or the contents tampered with after sealing. This makes the bottle the ideal solution for secure shipping or storage of samples, hazardous materials or high value contents.
The bottle is ideal for sealed sample shipments, ensuring your samples are safe and protected during transport.
Manufactured from HDPE and fitted with a liner screw cap, the bottle is not only strong but also liquid tight.
Suitable for liquids, powders, granules and pastes, the sealing bottle is versatile.
Catalog No. 50-000-25806 Supplier BURKLE INC Supplier No. 03221000
The sealable wide-neck bottle protects against unauthorized access or tampering. Sealing eyelets on the cap and bottle can be used to ensure that the bottle has not been opened or the contents tampered with after sealing. This makes the bottle the ideal solution for secure shipping or storage of samples, hazardous materials or high value contents.
The bottle is ideal for sealed sample shipments, ensuring your samples are safe and protected during transport.
Manufactured from HDPE and fitted with a liner screw cap, the bottle is not only strong but also liquid tight.
Suitable for liquids, powders, granules and pastes, the sealing bottle is versatile.