Learn More
BURKLE INC Wide-necked bottle, PP, round, 33.81 oz. (1000 ml), with cap
Catalog No. 5000025831
Encompass_Preferred
The wide-neck PP bottle is highly resistant to chemicals, and is shock and drop resistant. The bottle is also food safe and autoclavable.
The high transparency of PP makes it easy to see the contents of the bottle.
Catalog No. 50-000-25831 Supplier BURKLE INC Supplier No. 03191000
The wide-neck PP bottle is highly resistant to chemicals, and is shock and drop resistant. The bottle is also food safe and autoclavable.
The high transparency of PP makes it easy to see the contents of the bottle.