Learn More
BURKLE INC Wide-necked jerrycan, without thread, HDPE, 7.93 gal. (30l), withcap
Catalog No. 5000025603
Encompass_Preferred
Made of high-quality, transparent HDPE, the wide-neck canister is UV stabilized for lasting durability, even after prolonged sun exposure.
The 88 mm (3.46 in) wide filling opening ensures easy filling and effortless internal cleaning—ideal for sensitive or hard-to-clean substances.
The wide-mouth screw cap is attached with a safety strap, providing a secure, sealable closure. Grip grooves on the base ensure stable, safe handling during transport, filling, or emptying.
The canister includes a small venting and pouring spout (18 mm / 0.71 in diameter) for precise liquid removal and controlled venting during filling.
Catalog No. 50-000-25603 Supplier BURKLE INC Supplier No. 04310030
Made of high-quality, transparent HDPE, the wide-neck canister is UV stabilized for lasting durability, even after prolonged sun exposure.
The 88 mm (3.46 in) wide filling opening ensures easy filling and effortless internal cleaning—ideal for sensitive or hard-to-clean substances.
The wide-mouth screw cap is attached with a safety strap, providing a secure, sealable closure. Grip grooves on the base ensure stable, safe handling during transport, filling, or emptying.
The canister includes a small venting and pouring spout (18 mm / 0.71 in diameter) for precise liquid removal and controlled venting during filling.