Learn More
BURKLE INC Wide-necked jerrycan, with thread, HDPE, 2.91 gal. (11l), with cap
Supplier:  BURKLE INC 04311010
Encompass_Preferred
Made of premium, transparent HDPE, the UV-stabilized wide-neck canister offers long-lasting durability, even after extended sun exposure. Its extra-wide 88 mm opening allows easy filling and thorough internal cleaning—ideal for sensitive or hard-to-clean contents. The wide-mouth screw cap, secured with a safety strap, ensures a tight, reliable seal. Grip grooves at the base provide secure handling during transport, filling, or pouring. A built-in 0.71 in. (18 mm) venting and pouring spout allows controlled dispensing of small volumes and aids venting during filling. Positioned just above the base, the G 3/4" male spout enables attachment of a spigot or accessory for spill-free, accurate liquid dispensing. Designed for safety, convenience, and versatility, this canister is perfect for industrial, laboratory, and storage applications requiring clean and precise liquid handling.
Catalog No. 50-000-25551