Learn More
Hach Company Wide Range pH Indicator Solution, 100 mL MDB
Catalog No. 501995817 Shop All Hach Company Products
Encompass
For visual determination of pH, using test kits. pH 4-10 100 mL Marked Dropping Bottle
Catalog No. 50-199-5817 Supplier Hach Company Supplier No. 2329332
For visual determination of pH, using test kits. pH 4-10 100 mL Marked Dropping Bottle