Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Wide Range pH Test Kit
SDPSCI_ED

Catalog No. S96703 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96703 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96703 Supplier LaMotte™ Supplier No. 585801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Octa-Slide 2 Comparator, graduated test tubes and a bottle of indicator

This kit tests for pH using the Octa-Slide 2 Comparator with two color bars. Covers a wide pH range from 3.0 - 10.5 pH.

Simply fill the tube to the mark with sample water, add several drops of indicator and compare the resulting color against the permanent color standards.

  • Range: 3.0 to 10.5
  • Sensitivity: 0.5

Order Info

100 tests.

Specifications

Grade 3+
Product Type Kit, pH Test
Includes Octa-Slide 2 Comparator, graduated test tubes and a bottle of wide range indicator
For Use With (Application) Tests pH for drinking water, environmental education, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.