Learn More
FUJIFILM Biosciences WIDE-VIEW TM Prestained Protein Size Marker Ⅲ, 500uL
Catalog No. NC0357136
Encompass_Preferred
A stained protein size marker. There are twelve recombinant proteins contained, and they covalently bind to blue,red and green chromophores. A band for 75 kDa shows red, 25 kDa shows green and other bands exhibit blue color. This item should be kept at -20 Degrees Celcius.
Catalog No. NC0357136 Supplier FUJIFILM Biosciences Supplier No. 23002461
A stained protein size marker. There are twelve recombinant proteins contained, and they covalently bind to blue,red and green chromophores. A band for 75 kDa shows red, 25 kDa shows green and other bands exhibit blue color. This item should be kept at -20 Degrees Celcius.