Learn More
FUJIFILM Biosciences WIDE-VIEW TM Prestained Protein Size Marker Ⅲ
Catalog No. NC1466578
Encompass_Preferred
A stained protein size marker. There are twelve recombinant proteins contained, and they covalently bind to blue,red and green chromophores. A band for 75 kDa shows red, 25 kDa shows green and other bands exhibit blue color.
Catalog No. NC1466578 Supplier FUJIFILM Biosciences Supplier No. 23602463
A stained protein size marker. There are twelve recombinant proteins contained, and they covalently bind to blue,red and green chromophores. A band for 75 kDa shows red, 25 kDa shows green and other bands exhibit blue color.