Learn More
Perkin Elmer US LLC Wide-Bore Column Glass Liner
Catalog No. 502111510
Encompass_Preferred
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples. Inlet liners for splitless injection are generally designed as straight tubes, although new designs such as the gooseneck will help contain the sample in the injector. Packing these liners with wool will also help trap non-volatile residue and prevent column contamination. Compatible with the 480, 580, 680 and older Clarus GC models. Not compatible with Clarus 590/690 capillary injector.
Catalog No. 50-211-1510 Supplier Perkin Elmer US LLC Supplier No. N6101375
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples. Inlet liners for splitless injection are generally designed as straight tubes, although new designs such as the gooseneck will help contain the sample in the injector. Packing these liners with wool will also help trap non-volatile residue and prevent column contamination. Compatible with the 480, 580, 680 and older Clarus GC models. Not compatible with Clarus 590/690 capillary injector.