Learn More
Bachem 6-Carboxy-fluorescein
SDP
Catalog No. 501947389
Encompass_Preferred
6-Carboxy-fluorescein, 3301-79-9, C21H12O7, Mr 376.32, 500mg. 6-FAM, 2-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-terephthalic acid. Widely used green-fluorescent labeling dye.
Catalog No. 50-194-7389 Supplier Bachem Supplier No. 4045142.0500
6-Carboxy-fluorescein, 3301-79-9, C21H12O7, Mr 376.32, 500mg. 6-FAM, 2-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-terephthalic acid. Widely used green-fluorescent labeling dye.