Learn More
Electron Microscopy Sciences Shallow Wide Mouth Flasks, Size: 1000 ml. I.D.: 5.375. O.D.: 7. Height: 4.125., 1/EA
SDP
Catalog No. 502090676
Encompass_Preferred
Thermo-Flask features borosilicate glass inner liners which have been evacuate to 1x10-5mm of Hg to provide maximum coolant retention and safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2. Shallow Wide Mouth fasks have a dish-shaped design and are ideal for bench top use with crushed ice, dry- ice, or alcohol. Also great as a hot liquid container. Stainless steel jacket and cushioned base (lids not included) will provide years of trouble-free use. They are excellent for use as a cold trap. Size: 1000 ml. I.D.: 5.375 . O.D.: 7 . Height: 4.125.
Catalog No. 50-209-0676 Supplier Electron Microscopy Sciences Supplier No. 6189210
Thermo-Flask features borosilicate glass inner liners which have been evacuate to 1x10-5mm of Hg to provide maximum coolant retention and safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2. Shallow Wide Mouth fasks have a dish-shaped design and are ideal for bench top use with crushed ice, dry- ice, or alcohol. Also great as a hot liquid container. Stainless steel jacket and cushioned base (lids not included) will provide years of trouble-free use. They are excellent for use as a cold trap. Size: 1000 ml. I.D.: 5.375 . O.D.: 7 . Height: 4.125.