Learn More
Electron Microscopy Sciences Shallow Wide Mouth Flasks, 1900 ml, 6 I.D., 7.25 O.D., 5.5 height
SDP
Catalog No. 501899059
Encompass_Preferred
Thermo-Flask features borosilicate glass inner liners which have been evacuate to 1x10-5mm of Hg to provide maximum coolant retention and safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2. Shallow Wide Mouth fasks have a dish-shaped design and are ideal for bench top use with crushed ice, dry- ice, or alcohol. Also great as a hot liquid container. Stainless steel jacket and cushioned base (lids not included) will provide years of trouble-free use. They are excellent for use as a cold trap.
Size: 1900 ml
I.D.: 6
O.D.: 7.25
Height: 5.5
Catalog No. 50-189-9059 Supplier Electron Microscopy Sciences Supplier No. 6189219
Thermo-Flask features borosilicate glass inner liners which have been evacuate to 1x10-5mm of Hg to provide maximum coolant retention and safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2. Shallow Wide Mouth fasks have a dish-shaped design and are ideal for bench top use with crushed ice, dry- ice, or alcohol. Also great as a hot liquid container. Stainless steel jacket and cushioned base (lids not included) will provide years of trouble-free use. They are excellent for use as a cold trap.
Size: 1900 ml
I.D.: 6
O.D.: 7.25
Height: 5.5