Learn More
Waters Corp Method Scouting Kit, Reversed Phase, 3.5 μm
Catalog No. 502391763 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit contains a suite of columns chosen for general-purpose reversed-phase method development and scouting, offering diverse compound retention and selectivity.
  • Contains columns for general-purpose reversed-phase method development.
  • Columns selected for diverse compound retention and selectivity.
  • Includes 4.6 mm x 100 mm, 3.5 μm columns.
  • Kit includes columns such as BEH C18, BEH Shield RP18, CSH Fluoro-Phenyl, HSS Cyano, HSS C18 SB, and HSS T3.
  • Six units per package.
Catalog No. 50-239-1763 Supplier Waters Corp Supplier No. 176003362
Add to Cart
Add to Cart
Request a Quote
This kit contains a suite of columns chosen for general-purpose reversed-phase method development and scouting, offering diverse compound retention and selectivity.
  • Contains columns for general-purpose reversed-phase method development.
  • Columns selected for diverse compound retention and selectivity.
  • Includes 4.6 mm x 100 mm, 3.5 μm columns.
  • Kit includes columns such as BEH C18, BEH Shield RP18, CSH Fluoro-Phenyl, HSS Cyano, HSS C18 SB, and HSS T3.
  • Six units per package.