Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC WIF-1 Human HEK293 250ug
SDP

Catalog No. 5000197021
Encompass_Preferred

WIF-1 Protein, Human (HEK293, His) is a lyophilized recombinant human Wnt inhibitory factor-1, expressed in HEK293 cells with a C-terminal 6xHis tag. It is intended for research on WNT signaling and is supplied with certificate of analysis-verified purity, low endotoxin, and storage and reconstitution guidance for extended stability.

  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg).
  • Molecular weight approximately 42-48 kDa.
  • Supplied as lyophilized powder for stability.
  • Expression system HEK293 supports mammalian post-translational modifications.
  • Reconstitute at ≥100 μg/mL and add carrier protein for long-term storage.

Catalog No. 50-001-97021 Supplier Medchemexpress LLC Supplier No. HYP70982250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WIF-1 Protein, Human (HEK293, His) is a lyophilized recombinant human Wnt inhibitory factor-1, expressed in HEK293 cells with a C-terminal 6xHis tag. It is intended for research on WNT signaling and is supplied with certificate of analysis-verified purity, low endotoxin, and storage and reconstitution guidance for extended stability.

  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg).
  • Molecular weight approximately 42-48 kDa.
  • Supplied as lyophilized powder for stability.
  • Expression system HEK293 supports mammalian post-translational modifications.
  • Reconstitute at ≥100 μg/mL and add carrier protein for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.