Learn More
Lascar Electronics Inc High Accuracy 21CFR WiFi Temp & RH Data Logger
SDP
Catalog No. NC1856659
Encompass_Preferred
This high accuracy 21CFR WiFi-enabled temperature, humidity and dew point data logger can remotely monitor an environment over a -20 to +60°C (-4 to +140°­­F) and 0 to 100% RH measurement range.
Catalog No. NC1856659 Supplier Lascar Electronics Inc Supplier No. ELWIFI21CFRTH+
This high accuracy 21CFR WiFi-enabled temperature, humidity and dew point data logger can remotely monitor an environment over a -20 to +60°C (-4 to +140°­­F) and 0 to 100% RH measurement range.