Learn More
Lascar Electronics Inc WiFi Dual Channel Thermocouple Data Logge
SDP
Catalog No. NC1856633
Encompass_Preferred
his WiFi-enabled dual channel data logger can remotely monitor an environment using two inter-changeable J, K or T type thermocouples. Supplied with two K Type probes, its measurement range out of the box measures from 0 to 200°C (32 to 392°F).
Catalog No. NC1856633 Supplier Lascar Electronics Inc Supplier No. ELWIFIDTC
his WiFi-enabled dual channel data logger can remotely monitor an environment using two inter-changeable J, K or T type thermocouples. Supplied with two K Type probes, its measurement range out of the box measures from 0 to 200°C (32 to 392°F).