Learn More
Lascar Electronics Inc Dual Channel High Accuracy Thermistor WiFi
SDP
Catalog No. NC1856746
Encompass_Preferred
This WiFi-enabled dual channel data logger can remotely monitor an environment using two high accuracy thermistor probes. Supplied with two Type A probes, its measurement range out of the box measures from -40 to +125°C (-40 to +257°­­F). Data is uploaded periodically using a standard WiFi network to the EasyLog Cloud or a single host PC. EasyLog Cloud gives access to each data logger, and data collected from any internet-enabled device. Change data logger settings remotely, receive email alerts of alarm conditions, link sensors from multiple sites into one account and assign multiple user privileges.The device will store data internally if it loses WiFi connection and automatically uploads it to the Cloud once reconnected.Use the EasyLog Cloud Apps for Android and Apple to easily setup your device and access your data anytime, anywhere.
Catalog No. NC1856746 Supplier Lascar Electronics Inc Supplier No. ELWIFIDTP+
This WiFi-enabled dual channel data logger can remotely monitor an environment using two high accuracy thermistor probes. Supplied with two Type A probes, its measurement range out of the box measures from -40 to +125°C (-40 to +257°­­F). Data is uploaded periodically using a standard WiFi network to the EasyLog Cloud or a single host PC. EasyLog Cloud gives access to each data logger, and data collected from any internet-enabled device. Change data logger settings remotely, receive email alerts of alarm conditions, link sensors from multiple sites into one account and assign multiple user privileges.The device will store data internally if it loses WiFi connection and automatically uploads it to the Cloud once reconnected.Use the EasyLog Cloud Apps for Android and Apple to easily setup your device and access your data anytime, anywhere.