Learn More
Lascar Electronics Inc WiFi Thermistor Probe Data Logging Sensor
SDP
Catalog No. NC1856743
Encompass_Preferred
This high accuracy WiFi-enabled temperature data logging sensor can remotely monitor an environment over a -40 to 125°C (-40 to 257°­­F) measurement range. Data is uploaded periodically using a standard WiFi network to the EasyLog Cloud or a single host PC. Supplied with thermistor probe and mounting clip.EasyLog Cloud gives access to each data logger, and data collected from any internet-enabled device. Change data logger settings remotely, receive email alerts of alarm conditions, link sensors from multiple sites into one account and assign multiple user privileges.The device will store data internally if it loses WiFi connection and automatically uploads it to the Cloud once reconnected.Use the EasyLog Cloud Apps for Android and Apple to easily setup your device and access your data anytime, anywhere.
Catalog No. NC1856743 Supplier Lascar Electronics Inc Supplier No. ELWIFITP
This high accuracy WiFi-enabled temperature data logging sensor can remotely monitor an environment over a -40 to 125°C (-40 to 257°­­F) measurement range. Data is uploaded periodically using a standard WiFi network to the EasyLog Cloud or a single host PC. Supplied with thermistor probe and mounting clip.EasyLog Cloud gives access to each data logger, and data collected from any internet-enabled device. Change data logger settings remotely, receive email alerts of alarm conditions, link sensors from multiple sites into one account and assign multiple user privileges.The device will store data internally if it loses WiFi connection and automatically uploads it to the Cloud once reconnected.Use the EasyLog Cloud Apps for Android and Apple to easily setup your device and access your data anytime, anywhere.