Learn More
Thermo Scientific™ Wig-L-Bug™ Mixing/Grinding Mill Accessory, Stainless-Steel Vial
Catalog No. AA42629KT
Change view
Click to view available options
For Use With (Equipment):
Wig-L-Bug Mixing/Grinding Mill
Catalog No. For Use With (Equipment)
AA42629KT Wig-L-Bug Mixing/Grinding Mill
1 options
Catalog No. AA42629KT Supplier Thermo Scientific™ Supplier No. 42629KT
For use with Wig-L-Bug Mixing/Grinding Mill (AA42627KT)
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Stainless-steel vial, 1 in. long x 1/2 in. diameter (2.54 x 1.27cm)
  • With stainless-steel ball pestle, 1/4 in. diameter (0.635cm)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stainless-Steel Vial
For Use With (Equipment) Wig-L-Bug Mixing/Grinding Mill