Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wijs solution, Volumetric, 0.1 M ICl, Solstice

Catalog No. 60046523 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-523 4 x 2.5 L
60-017-68 1 L
60-017-69 2.5 L
60-046-524 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-523 Supplier Solstice Supplier No. 350714X2.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 0.1 M ICl

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7 , 7790-99-0
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 24640
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Quantity 4 x 2.5 L
Linear Formula ICl
MDL Number MFCD00036152
UN Number UN2920
Flash Point 40°C (104°F)
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
Formula Weight 162.36
Chemical Name or Material Wijs solution
Hazard Statement P280-P305 + P351 + P338-P310
DOTInformation 3(8) / PGII
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.