Learn More
TARGETMOL CHEMICALS INC WIKI4 25MG
SDP
Catalog No. 502580399
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WIKI4 is a potent inhibitor of Wnt/(beta)-catenin signaling and tankyrase 2 (TNKS2). purity: 99%
Catalog No. 50-258-0399 Supplier TARGETMOL CHEMICALS INC Supplier No. T306225MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WIKI4 is a potent inhibitor of Wnt/(beta)-catenin signaling and tankyrase 2 (TNKS2). purity: 99%