Learn More
IBL America sAPPβ-Wild Type Anti-Human Rabbit IgG Affinity Purify
Supplier:  IBL America 18957
Encompass_Preferred
For research use only, not for use in diagnostic procedures.This antibody can detect sAPP-Wild type which is cleavaged by β-secretase. Non-crossreacts with sAPPα and full-length APP.
Catalog No. NC1010001