Learn More
WILDCAT WHOLESALE 2" Cardboard White Freezer Boxes w/81-Place Divider 5x5 inches
Catalog No. NC1616798
Encompass_Preferred
This item has a minimum order quantity of 72 per supplier requirements2" Cardboard White Freezer Boxes w/81-Place Divider w/Drain Slots. Size (ID): 4-15/16" x 4-15/16" x 1-7/8" (OD): 5" x 5" x 2". Cover is 1-1/4" Deep. Perfect for Biological Sample Storage or vials in various storage racks. White in and White out. 81-Place Divider Arranged 9x9.
Catalog No. NC1616798 Supplier WILDCAT WHOLESALE Supplier No. BOX2DIV
This item has a minimum order quantity of 72 per supplier requirements2" Cardboard White Freezer Boxes w/81-Place Divider w/Drain Slots. Size (ID): 4-15/16" x 4-15/16" x 1-7/8" (OD): 5" x 5" x 2". Cover is 1-1/4" Deep. Perfect for Biological Sample Storage or vials in various storage racks. White in and White out. 81-Place Divider Arranged 9x9.