Learn More
WILDCAT WHOLESALE Racks Boxes Labeling and Tape, Boxes, Cryogenic and Freezer Boxes, WILDCAT DIVIDERS FOR CRYO/FREEZER BOX, 81-Place (9x9), Fits 2"" and 3"" boxes, White, Cell: 17/32"" x 17/32""(13mm x 13mm)
Catalog No. NC1616881
Encompass_Preferred
This item has a minimum order quantity of 50 per supplier requirements
Racks Boxes Labeling and Tape, Boxes, Cryogenic and Freezer Boxes, WILDCAT DIVIDERS FOR CRYO/FREEZER BOX, 81-Place (9x9), Fits 2" and 3" boxes, White, Cell: 17/32" x 17/32"(13mm x 13mm)
Catalog No. NC1616881 Supplier WILDCAT WHOLESALE Supplier No. DIV81
This item has a minimum order quantity of 50 per supplier requirements
Racks Boxes Labeling and Tape, Boxes, Cryogenic and Freezer Boxes, WILDCAT DIVIDERS FOR CRYO/FREEZER BOX, 81-Place (9x9), Fits 2" and 3" boxes, White, Cell: 17/32" x 17/32"(13mm x 13mm)