Learn More
WILDCAT WHOLESALE WILDCAT™ Dividers For Cryo/Freezer Box, 36-Place (6x6), White, Cell: 25/32"""" x 25/32""""(20mm x 20mm)
Catalog No. NC1616890
Encompass_Preferred
This item has a minimum order quantity of 50 per supplier requirements
WILDCAT™ Dividers For Cryo/Freezer Box, 36-Place (6x6), Fits 2" and 3" boxes, White, Cell: 25/32" x 25/32"(20mm x 20mm)
Catalog No. NC1616890 Supplier WILDCAT WHOLESALE Supplier No. DIV36
This item has a minimum order quantity of 50 per supplier requirements
WILDCAT™ Dividers For Cryo/Freezer Box, 36-Place (6x6), Fits 2" and 3" boxes, White, Cell: 25/32" x 25/32"(20mm x 20mm)