Learn More
Med Vet International Miltex Vantage Wilde Ear Forceps, 4.75in, Angular
Supplier:  Med Vet International MILV919350
Encompass_Preferred
The Miltex Vantage V919-350 Wilde Ear Forceps are made of mid-grade stainless steel and have a length of 4.75in. They have a non-locking bayonet handle and straight serrated blades. Used in a variety of ENT procedures they are ideal for nasal packing, grabbing delicate soft tissue, and grabbing ear tubes.
Catalog No. 50-255-9467