Learn More
Drager Wildland Respiratory Protect Kit
Catalog No. 17010220 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-220 Large
17-010-218 Small
17-010-219 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-010-220 Supplier Drager Supplier No. NA10754
Add to Cart
Add to Cart
The Drager X-plore 3500 Wildland Respiratory Kit helps protect wearers against smoke particles from wildfires.
  • WILDLAND RESPIRATORY KIT: This half-mask respirator kit is designed for those living and working in the vicinity of wildfires or bushfires. It comes with a practical carrying case as well as a P100 particle filter.
  • COMFORT FOR EXTREME SITUATIONS: Thanks to the DragerFlex material the mask body is extremely comfortable and adjusts extremely good to every face. Thus, the mask protects firemen, first responders, paramedics and civilians even over a long periods of time in the most comfortable way.
  • HIGH PROTECTION: The reusable P100 filter keeps you protected at all times and can easily be replaced if needed. Both mask and filter are NIOSH-approved and protect against particulates, dust and ash from wildfires.
  • RELIABLE EQUIPMENT: The kit contains the Drager half mask X-plore 3500, 2 P100 particle filters, a carrying bag and cleaning wipes for easy maintenance.
Specifications
Aerosol Type P - Oil-Proof
Color Black
Design Type Cup/Molded
Disposable Reusable
Filter Type Particulate
Material EPDM
Product Type General Purpose Particulate Respirator
Size Large
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile