Learn More
TA Instruments Wiley NIST Mass Spectrometer Library, Identification of Unknown Samples, Comparison to Known Material Ionization Cracking Patterns
Catalog No. 5000580845 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Wiley/NIST Mass Spectrometer Library is used for identification of unknown samples. The software compares the mass spectrometry ionization cracking pattern of unknown samples with those of known individual materials.
Catalog No. 50-005-80845 Supplier TA Instruments Supplier No. 271003.001
Waters TA Instruments Wiley/NIST Mass Spectrometer Library is used for identification of unknown samples. The software compares the mass spectrometry ionization cracking pattern of unknown samples with those of known individual materials.