Learn More
MSC Wiley X Romer 3 Sunglasses, Matte Black Frame, 1 ea
Catalog No. 5000676611
Encompass
The Wiley X Romer 3 features a sleek design, compatible with night vision goggles (NVG), offering maximum peripheral vision with its Wrapback™ design. It includes Slot™ technology for changeable lenses with Foil™ anti-fog and T-shell™ scratch-resistant coatings. Shatterproof selenite polycarbonate lenses provide 100% UVA/UVB protection and distortion-free clarity.
  • Shatterproof selenite™ polycarbonate lenses with 100% UVA and UVB protection
  • T-shell™ coating resists scratching
  • Foil™ coating provides anti-fog protection
  • Sleek design, night vision goggle (NVG) compatible
  • Wrapback™ design for maximum peripheral vision
  • Slot™ technology for changeable lens function
  • Rx ready
  • Includes light rust, smoke grey, and clear lenses
  • Protective storage case and microfiber cleaning cloth
Catalog No. 50-006-76611 Supplier MSC Supplier No. 41681917
The Wiley X Romer 3 features a sleek design, compatible with night vision goggles (NVG), offering maximum peripheral vision with its Wrapback™ design. It includes Slot™ technology for changeable lenses with Foil™ anti-fog and T-shell™ scratch-resistant coatings. Shatterproof selenite polycarbonate lenses provide 100% UVA/UVB protection and distortion-free clarity.
  • Shatterproof selenite™ polycarbonate lenses with 100% UVA and UVB protection
  • T-shell™ coating resists scratching
  • Foil™ coating provides anti-fog protection
  • Sleek design, night vision goggle (NVG) compatible
  • Wrapback™ design for maximum peripheral vision
  • Slot™ technology for changeable lens function
  • Rx ready
  • Includes light rust, smoke grey, and clear lenses
  • Protective storage case and microfiber cleaning cloth